siRNA Details
Gene Symbol: Snhg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027059.1 | 2 | 585 | |||
GenBank mRNA | AK012687.1 | - | 2 | 579 | ||
AK035703.1 | - | 1 | 976 | |||
AK171830.1 | - | 1 | 2141 | |||
BC037520.1 | - | 2 | 505 |
