siRNA Details
Gene Symbol: Snhg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027059.1 | 1 | 390 | |||
GenBank mRNA | AK012687.1 | - | 1 | 384 | ||
AK171830.1 | - | 1 | 372 | |||
BC037520.1 | - | 1 | 310 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snhg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027059.1 | 1 | 390 | |||
GenBank mRNA | AK012687.1 | - | 1 | 384 | ||
AK171830.1 | - | 1 | 372 | |||
BC037520.1 | - | 1 | 310 |