siRNA Details
Gene Symbol: 4930481A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015545.2 | 1 | 184 | |||
NR_027938.2 | 1 | 184 | ||||
GenBank mRNA | AK015604.1 | - | 1 | 185 | ||
AK039982.1 | - | 1 | 171 | |||
BC065793.1 | - | 1 | 114 | |||
BC085508.1 | - | 1 | 125 | |||
BM937611.1 | - | 1 | 113 |
