siRNA Details
Gene Symbol: Snhg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029468.1 | 1 | 117 | |||
GenBank mRNA | AK009175.1 | - | 1 | 121 | ||
BY130275.1 | - | 1 | 118 | |||
CB201672.1 | - | 1 | 102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snhg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029468.1 | 1 | 117 | |||
GenBank mRNA | AK009175.1 | - | 1 | 121 | ||
BY130275.1 | - | 1 | 118 | |||
CB201672.1 | - | 1 | 102 |