siRNA Details
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038656.1 | - | 1 | 66 | ||
BC037216.1 | - | 1 | 158 | 5361681 | ||
BY130176.1 | - | 1 | 237 | |||
CJ059997.1 | - | 1 | 186 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038656.1 | - | 1 | 66 | ||
BC037216.1 | - | 1 | 158 | 5361681 | ||
BY130176.1 | - | 1 | 237 | |||
CJ059997.1 | - | 1 | 186 |