siRNA Details
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037216.1 | - | 1 | 34 | 5361681 | |
BY130176.1 | - | 1 | 113 | |||
CJ059997.1 | - | 1 | 62 |

More Information
Other Species Targeted

