siRNA Details
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BY130176.1 | - | 1 | 40 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ054519.1 | - | 1 | 22 | Chr. 6 - 80018895 - 80021749 [+] on Build GRCm38 |
