siRNA Details
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033197.1 | 1 | 1145 | |||
GenBank mRNA | AK006196.1 | - | 1 | 934 | ||
AK016947.1 | - | 1 | 1146 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033197.1 | 1 | 1145 | |||
GenBank mRNA | AK006196.1 | - | 1 | 934 | ||
AK016947.1 | - | 1 | 1146 |