siRNA Details
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033197.1 | 1 | 572 | |||
GenBank mRNA | AK006089.1 | - | 1 | 386 | ||
AK006196.1 | - | 1 | 361 | |||
AK006985.1 | - | 1 | 383 | |||
AK016947.1 | - | 1 | 573 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716627.1 | - | 1 | 573 | Chr. 15 - 74709161 - 74710046 [+] on Build GRCm38 |
