siRNA Details
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033197.1 | 1 | 523 | |||
GenBank mRNA | AK006089.1 | - | 1 | 337 | ||
AK006196.1 | - | 1 | 312 | |||
AK006985.1 | - | 1 | 334 | |||
AK016947.1 | - | 1 | 524 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716627.1 | - | 1 | 524 | Chr. 15 - 74709161 - 74710046 [+] on Build GRCm38 |
