siRNA Details
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033197.1 | 1 | 698 | |||
GenBank mRNA | AK006089.1 | - | 1 | 512 | ||
AK006196.1 | - | 1 | 487 | |||
AK006985.1 | - | 1 | 509 | |||
AK016947.1 | - | 1 | 699 |
