siRNA Details
Gene Symbol: 4930471G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015571.1 | 1, 2 | 101 | |||
GenBank mRNA | AK015550.1 | - | 1, 2 | 111 | ||
BC048509.1 | - | 1, 2 | 99 | |||
BC061192.1 | - | 1, 2 | 87 | |||
BC100467.1 | - | 1, 2 | 101 |
