siRNA Details
Gene Symbol: 4930471G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015571.1 | 2 | 198 | |||
GenBank mRNA | AK015550.1 | - | 2 | 208 | ||
AK018831.1 | - | 2 | 218 | |||
BC048509.1 | - | 2 | 196 | |||
BC061192.1 | - | 2 | 184 | |||
BC100467.1 | - | 2 | 198 |
