siRNA Details
Gene Symbol: Dnm3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002870.2 | 1 | 3081 | |||
GenBank mRNA | AK053654.1 | - | 1 | 1865 | ||
AK164873.1 | - | 1 | 909 | |||
AY751519.1 | - | 1 | 3041 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnm3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002870.2 | 1 | 3081 | |||
GenBank mRNA | AK053654.1 | - | 1 | 1865 | ||
AK164873.1 | - | 1 | 909 | |||
AY751519.1 | - | 1 | 3041 |