siRNA Details
Gene Symbol: Gm4926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028066.1 | 1 | 153 | |||
GenBank mRNA | AK079066.1 | - | 1 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625816.1 | - | 1 | 151 | Chr. 11 - 46637496 - 46645004 [-] on Build GRCm38 |
