siRNA Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015456.1 | 1 | 222 | |||
GenBank mRNA | AK035508.1 | - | 1 | 223 | ||
AK035645.1 | - | 1 | 223 | |||
AK039454.1 | - | 1060 | ||||
AK080843.1 | - | 1551 | ||||
BC070450.1 | - | 48 |
