siRNA Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015456.1 | 3 | 2043 | |||
GenBank mRNA | AK035508.1 | - | 3 | 2044 | ||
AK035645.1 | - | 3 | 2044 | |||
AK078094.1 | - | 3 | 1689 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015456.1 | 3 | 2043 | |||
GenBank mRNA | AK035508.1 | - | 3 | 2044 | ||
AK035645.1 | - | 3 | 2044 | |||
AK078094.1 | - | 3 | 1689 |