siRNA Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015456.1 | 1 | 315 | |||
GenBank mRNA | AK035508.1 | - | 1 | 316 | ||
AK035645.1 | - | 1 | 316 | |||
AK039454.1 | - | 1153 | ||||
AK080843.1 | - | 1644 | ||||
BC070450.1 | - | 141 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD803437.1 | - | 1 | 141 | Chr. 7 - 59971900 - 59974257 [-] on Build GRCm38 |
