siRNA Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015456.1 | 3 | 1616 | |||
GenBank mRNA | AK035508.1 | - | 3 | 1617 | ||
AK035645.1 | - | 3 | 1617 | |||
AK078094.1 | - | 3 | 1262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB236614.1 | - | 3 | 806 | Chr. 7 - 59970092 - 59973617 [-] on Build GRCm38 |
