siRNA Details
Gene Symbol: Snhg18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038186.1 | 2 | 418 | |||
GenBank mRNA | AK002304.1 | - | 2 | 419 | ||
AK007520.1 | - | 2 | 175 | |||
AK007719.1 | - | 2 | 190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snhg18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038186.1 | 2 | 418 | |||
GenBank mRNA | AK002304.1 | - | 2 | 419 | ||
AK007520.1 | - | 2 | 175 | |||
AK007719.1 | - | 2 | 190 |