siRNA Details
Gene Symbol: 2900079G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015468.1 | 2 | 680 | |||
NR_033431.1 | 2 | 558 | ||||
GenBank mRNA | AK013806.1 | - | 2 | 559 | ||
AK038201.1 | - | 1 | 1317 | |||
BC056188.1 | - | 2 | 680 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX238067.1 | - | 2 | 540 | Chr. 9 - 112234295 - 112235755 [+] on Build GRCm38 |
