siRNA Details
Gene Symbol: 2900079G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015468.1 | 1 | 33 | |||
GenBank mRNA | AK038201.1 | - | 1 | 172 | ||
BC056188.1 | - | 1 | 33 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ445424.1 | - | 1 | 33 | Chr. 9 - 112234381 - 112235576 [+] on Build GRCm38 |
