siRNA Details
Gene Symbol: 2900079G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015468.1 | 2 | 950 | |||
NR_033431.1 | 2 | 828 | ||||
GenBank mRNA | AK013806.1 | - | 2 | 829 | ||
AK038201.1 | - | 1 | 1587 | |||
BC056188.1 | - | 2 | 950 |
