siRNA Details
Gene Symbol: D730001G18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027836.1 | 3 | 988 | |||
NR_027837.1 | 2 | 889 | ||||
GenBank mRNA | AK021322.1 | - | 2 | 890 | ||
AK037167.1 | - | 2 | 889 | |||
BC055822.1 | - | 3 | 953 | |||
BC147771.1 | - | 1 | 769 | |||
BC147774.1 | - | 1 | 769 |
