siRNA Details
Gene Symbol: 4930511M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015494.1 | 3 | 494 | |||
NR_027944.1 | 2 | 339 | ||||
NR_027945.1 | 3 | 324 | ||||
GenBank mRNA | AK015760.1 | - | 3 | 337 | ||
AK144587.1 | - | 3 | 462 | |||
AK144776.1 | - | 2 | 340 | |||
AK165835.1 | - | 146 | ||||
AK165876.1 | - | 3 | 495 |
