siRNA Details
Gene Symbol: Zfp783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027963.1 | 6 | 4389 | |||
GenBank mRNA | AK220535.1 | - | 6 | 4261 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027963.1 | 6 | 4389 | |||
GenBank mRNA | AK220535.1 | - | 6 | 4261 |