siRNA Details
Gene Symbol: Gm10941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026944.1 | 1 | 127 | |||
GenBank mRNA | AK132904.1 | - | 1 | 128 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ047168.1 | - | 1 | 128 | Chr. 10 - 77257836 - 77259223 [-] on Build GRCm38 |
