siRNA Details
Gene Symbol: 9430076C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015553.2 | 5 | 470 | |||
GenBank mRNA | AK051275.1 | - | 5 | 470 | ||
AK166503.1 | - | 5 | 597 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB656461.1 | - | 5 | 470 | Chr. 6 - 53287295 - 53394843 [+] on Build GRCm38 |
