siRNA Details
Gene Symbol: 1700111N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033213.2 | 2, 3 | 170 | |||
GenBank mRNA | AK007172.1 | - | 2, 3 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF104396.1 | - | 2, 3 | 138 | Chr. X - 69979618 - 70143556 [-] on Build GRCm38 |
