siRNA Details
Gene Symbol: Gm4489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027637.1 | 3 | 445 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU698641.1 | - | 3 | 448 | Chr. 10 - 122119697 - 122130554 [+] on Build GRCm38 |
