siRNA Details
Gene Symbol: 2900092D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027891.1 | 1 | 2391 | |||
GenBank mRNA | AK038597.1 | - | 1 | 1582 | ||
AK053590.1 | - | 1 | 1571 | |||
AK163519.1 | - | 1 | 1576 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB174009.2 | - | 1 | 274 | Chr. 1 - 42702517 - 42703176 [+] on Build GRCm38 |
