siRNA Details
Gene Symbol: A330035P11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015586.2 | 4 | 2671 | |||
GenBank mRNA | AK039382.1 | - | 4 | 2672 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A330035P11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015586.2 | 4 | 2671 | |||
GenBank mRNA | AK039382.1 | - | 4 | 2672 |