siRNA Details
Gene Symbol: A330035P11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015586.2 | 1 | 368 | |||
GenBank mRNA | AK031460.1 | - | 1 | 311 | ||
AK032758.1 | - | 1 | 369 | |||
AK039382.1 | - | 1 | 369 | |||
AK049508.1 | - | 1 | 348 |
