siRNA Details
Gene Symbol: 1700009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015547.1 | 1 | 246 | |||
GenBank mRNA | AK005796.1 | - | 1 | 247 | ||
AK006749.1 | - | 1 | 231 | |||
AK015973.1 | - | 1 | 240 | |||
BC048578.1 | - | 1 | 91 | |||
BC061019.1 | - | 1 | 246 |
