siRNA Details
Gene Symbol: 1700009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015547.1 | 2 | 805 | |||
GenBank mRNA | AK005796.1 | - | 2 | 806 | ||
AK006749.1 | - | 2 | 790 | |||
BC061019.1 | - | 2 | 805 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015547.1 | 2 | 805 | |||
GenBank mRNA | AK005796.1 | - | 2 | 806 | ||
AK006749.1 | - | 2 | 790 | |||
BC061019.1 | - | 2 | 805 |