siRNA Details
Gene Symbol: 1700009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015547.1 | 2 | 747 | |||
GenBank mRNA | AK006749.1 | - | 2 | 732 | ||
BC048578.1 | - | 2 | 592 | |||
BC061019.1 | - | 2 | 747 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015547.1 | 2 | 747 | |||
GenBank mRNA | AK006749.1 | - | 2 | 732 | ||
BC048578.1 | - | 2 | 592 | |||
BC061019.1 | - | 2 | 747 |