siRNA Details
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003520.1 | 1 | 110 | |||
GenBank mRNA | AK149635.1 | - | 1 | 110 | ||
AK149792.1 | - | 1 | 110 | |||
AK150023.1 | - | 1 | 110 | |||
AK150236.1 | - | 1 | 110 |
