siRNA Details
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003520.1 | 9 | 2455 | |||
GenBank mRNA | AK076430.1 | - | 14 | 2726 | ||
AK150023.1 | - | 9 | 2454 | |||
AK150236.1 | - | 9 | 2454 | |||
M21039.1 | - | 11 | 2550 |
