siRNA Details
Gene Symbol: 4831440E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030700.1 | 1 | 924 | |||
GenBank mRNA | AK029244.1 | - | 1 | 780 | ||
AK087767.1 | - | 208 | ||||
AK149324.1 | - | 1 | 923 | |||
AK163107.1 | - | 1 | 551 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB666475.1 | - | 2 | 204 | Chr. 5 - 25499070 - 25501165 [+] on Build GRCm38 |
