siRNA Details
Gene Symbol: 1700020N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026924.1 | 1 | 41 | |||
GenBank mRNA | AK006182.1 | - | 1 | 42 | ||
AK006756.1 | - | 1 | 42 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700020N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026924.1 | 1 | 41 | |||
GenBank mRNA | AK006182.1 | - | 1 | 42 | ||
AK006756.1 | - | 1 | 42 |