siRNA Details
Gene Symbol: 5430402O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015581.1 | 1 | 330 | |||
NR_027892.1 | 1 | 330 | ||||
GenBank mRNA | AK014187.1 | - | 1 | 233 | ||
AK017262.1 | - | 1 | 235 | |||
AK088092.1 | - | 1 | 328 | |||
AK139045.1 | - | 1 | 331 |
