siRNA Details
Gene Symbol: 5430402O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015581.1 | 4 | 805 | |||
NR_027892.1 | 3 | 738 | ||||
GenBank mRNA | AK017262.1 | - | 4 | 710 | ||
AK088092.1 | - | 3 | 736 | |||
AK139045.1 | - | 3 | 740 |
