siRNA Details
Gene Symbol: 5430402O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015581.1 | 1 | 63 | |||
NR_027892.1 | 1 | 63 | ||||
GenBank mRNA | AK088092.1 | - | 1 | 61 | ||
AK139045.1 | - | 1 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY746058.1 | - | 1 | 61 | Chr. 6 - 50566658 - 50594707 [+] on Build GRCm38 |
