siRNA Details
Gene Symbol: 5430402O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015581.1 | 4 | 924 | |||
NR_027892.1 | 3 | 857 | ||||
GenBank mRNA | AK017262.1 | - | 4 | 829 | ||
AK139045.1 | - | 3 | 859 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5430402O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015581.1 | 4 | 924 | |||
NR_027892.1 | 3 | 857 | ||||
GenBank mRNA | AK017262.1 | - | 4 | 829 | ||
AK139045.1 | - | 3 | 859 |