siRNA Details
Gene Symbol: 5430402O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015581.1 | 4 | 774 | |||
NR_027892.1 | 3 | 707 | ||||
GenBank mRNA | AK017262.1 | - | 4 | 679 | ||
AK088092.1 | - | 3 | 705 | |||
AK139045.1 | - | 3 | 709 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY761904.1 | - | 3 | 761 | Chr. 6 - 50566687 - 50594865 [+] on Build GRCm38 |
