siRNA Details
Gene Symbol: 2010016I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033207.1 | 2 | 1444 | |||
GenBank mRNA | AK040034.1 | - | 1 | 1355 | ||
AK079963.1 | - | 2 | 745 | |||
AK080971.1 | - | 2 | 1445 | |||
AK140363.1 | - | 2 | 1445 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB215580.2 | - | 2 | 489 | Chr. 3 - 106482003 - 106483578 [-] on Build GRCm38 |
