siRNA Details
Gene Symbol: Mir10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029784.1 | 1 | 32 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NR_031814.
Gene Ontology Categories:
