siRNA Details
Gene Symbol: 1700031M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015496.1 | 2 | 108 | |||
GenBank mRNA | AK006585.1 | - | 2 | 109 | ||
AK015131.1 | - | 2 | 110 | |||
BC048654.1 | - | 2 | 108 | |||
BC099572.1 | - | 2 | 116 |
