siRNA Details
Gene Symbol: 2610306M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028298.1 | 1 | 776 | |||
GenBank mRNA | AK011997.1 | - | 1 | 777 | ||
AK015614.1 | - | 1 | 750 | |||
BC058130.1 | - | 1 | 489 | |||
BC096561.1 | - | 1 | 281 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711193.1 | - | 1 | 773 | Chr. 6 - 86848544 - 86849437 [-] on Build GRCm38 |
