siRNA Details
Gene Symbol: 2610306M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028298.1 | 1 | 953 | |||
GenBank mRNA | AK011997.1 | - | 1 | 954 | ||
BC058130.1 | - | 1 | 666 | |||
BC096561.1 | - | 1 | 458 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV127221.1 | - | 1 | 213 | Chr. 6 - 86848408 - 86848701 [-] on Build GRCm38 |
